siRNA Details
| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172662.3 | NP_766250.2 | 10 | 1418 | ||
| GenBank mRNA | AK054272.1 | - | 10 | 1419 | ||
| AK162289.1 | - | 2 | 205 | |||
| BC052069.1 | - | 7 | 983 | 5715781 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtdc1| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172662.3 | NP_766250.2 | 10 | 1418 | ||
| GenBank mRNA | AK054272.1 | - | 10 | 1419 | ||
| AK162289.1 | - | 2 | 205 | |||
| BC052069.1 | - | 7 | 983 | 5715781 | ||