siRNA Details
| Gene Symbol: Acad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172678.3 | NP_766266.3 | 15 | 1635 | ||
| GenBank mRNA | AK012773.1 | - | 2 | 160 | ||
| AK028571.1 | - | 14 | 2349 | |||
| AK031820.1 | - | 15 | 1567 | |||
| AK075984.1 | - | 15 | 1554 | |||
| AK088409.1 | - | 2 | 70 | |||
| AK145423.1 | - | 15 | 1543 | |||
| AK181172.1 | - | 5 | 409 | |||
| BC004737.1 | - | 8 | 1524 | |||
| BC031137.1 | - | 15 | 1571 | 4971848 | ||
| BC032213.1 | - | 15 | 1576 | 5323723 | ||
| BC033277.1 | - | 15 | 1556 | 4241041 | ||

Acad9
Genomic Map