siRNA Details
| Gene Symbol: Rsbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172684.2 | NP_766272.2 | 2 | 1396 | ||
| GenBank mRNA | AB101658.1 | - | 2 | 1398 | ||
| AK087685.1 | - | 1 | 362 | |||
| AK122577.1 | - | 2 | 931 | |||
| AK134093.1 | - | 2 | 397 | |||
| AK138603.1 | - | 2 | 940 | |||
| BC094667.1 | - | 2 | 921 | |||
| BC139213.1 | - | 2 | 1365 | 8862235 | ||
| BC139215.1 | - | 2 | 1365 | 8862237 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191710, XM_006233082.

Rsbn1
Genomic Map