siRNA Details
| Gene Symbol: N28178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172690.2 | NP_766278.1 | 10 | 2726 | ||
| GenBank mRNA | AK038528.1 | - | 9 | 2580 | ||
| AK122428.1 | - | 8 | 2649 | |||
| BC057092.1 | - | 10 | 2726 | 5703966 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

N28178| Gene Symbol: N28178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172690.2 | NP_766278.1 | 10 | 2726 | ||
| GenBank mRNA | AK038528.1 | - | 9 | 2580 | ||
| AK122428.1 | - | 8 | 2649 | |||
| BC057092.1 | - | 10 | 2726 | 5703966 | ||