siRNA Details
| Gene Symbol: Phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172705.2 | NP_766293.2 | 4 | 2575 | ||
| GenBank mRNA | AK077052.1 | - | 2 | 2091 | ||
| BC020286.1 | - | 2 | 1624 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf13| Gene Symbol: Phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172705.2 | NP_766293.2 | 4 | 2575 | ||
| GenBank mRNA | AK077052.1 | - | 2 | 2091 | ||
| BC020286.1 | - | 2 | 1624 | |||