siRNA Details
| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172708.3 | NP_766296.1 | 3 | 300 | ||
| GenBank mRNA | AB220919.1 | - | 4 | 346 | ||
| AK044445.1 | - | 3 | 301 | |||
| BC089590.1 | - | 4 | 419 | 6759981 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dok7| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172708.3 | NP_766296.1 | 3 | 300 | ||
| GenBank mRNA | AB220919.1 | - | 4 | 346 | ||
| AK044445.1 | - | 3 | 301 | |||
| BC089590.1 | - | 4 | 419 | 6759981 | ||