siRNA Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172719.2 | NP_766307.2 | 38, 39 | 4981 | ||
| GenBank mRNA | AB093217.1 | - | 14, 15 | 2174 | ||
| AK077013.1 | - | 2, 3 | 303 | |||
| AK090137.1 | - | 38, 39 | 4979 | |||
| AK147257.1 | - | 38, 39 | 4974 | |||
| BC056933.1 | - | 4, 5 | 613 | |||
| BC082548.1 | - | 25, 26 | 3759 | |||
| BC150735.1 | - | 38, 39 | 4984 | 9087646 | ||

Gcn1l1
Genomic Map