siRNA Details
| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205363.1 | NP_001192292.1 | 1, 2 | 192 | ||
| NM_001205365.1 | NP_001192294.1 | 1, 2 | 192 | |||
| NM_172732.3 | NP_766320.2 | 1, 2 | 192 | |||
| GenBank mRNA | AK036399.1 | - | 1, 2 | 193 | ||
| AK041288.1 | - | 1, 2 | 192 | |||
| BC087955.1 | - | 1, 2 | 191 | 30119997 | ||
| EU339277.1 | - | 1, 2 | 99 | |||
| EU339278.1 | - | 1, 2 | 99 | |||
| EU339280.1 | - | 1, 2 | 99 | |||
| EU339281.1 | - | 1, 2 | 99 | |||

Clec9a
Genomic Map