siRNA Details
| Gene Symbol: 4931406P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172741.2 | NP_766329.1 | 1 | 655 | ||
| GenBank mRNA | AK076994.1 | - | 1 | 656 | ||
| AK134699.1 | - | 2 | 1056 | |||
| BC060233.1 | - | 1 | 456 | 6852905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931406P16Rik| Gene Symbol: 4931406P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172741.2 | NP_766329.1 | 1 | 655 | ||
| GenBank mRNA | AK076994.1 | - | 1 | 656 | ||
| AK134699.1 | - | 2 | 1056 | |||
| BC060233.1 | - | 1 | 456 | 6852905 | ||