siRNA Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172772.2 | NP_766360.2 | 1, 2 | 910 | ||
| GenBank mRNA | AK035930.1 | - | 1, 2 | 911 | ||
| AK046403.1 | - | 1, 2 | 897 | |||
| AK046623.1 | - | 1, 2 | 884 | |||
| BC055816.1 | - | 1, 2 | 638 | |||
| BC058683.1 | - | 1, 2 | 868 | 6834871 | ||

Fam63b
Genomic Map