siRNA Details
| Gene Symbol: Slc22a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172776.2 | NP_766364.1 | 8 | 1495 | ||
| GenBank mRNA | AF536195.1 | - | 8 | 1567 | ||
| AF536196.1 | - | 9 | 1303 | |||
| AK052597.1 | - | 8 | 1496 | |||
| BC126952.1 | - | 9 | 1308 | 40090863 | ||

Slc22a29
Genomic Map