siRNA Details
| Gene Symbol: Il20ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172786.2 | NP_766374.1 | 3 | 339 | ||
| GenBank mRNA | AK054215.1 | - | 3 | 340 | ||
| BC056628.1 | - | 3 | 323 | 5033177 | ||
| EF592560.1 | - | 4 | 417 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il20ra| Gene Symbol: Il20ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172786.2 | NP_766374.1 | 3 | 339 | ||
| GenBank mRNA | AK054215.1 | - | 3 | 340 | ||
| BC056628.1 | - | 3 | 323 | 5033177 | ||
| EF592560.1 | - | 4 | 417 | |||