siRNA Details
| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 2, 3 | 317 | ||
| GenBank mRNA | AK030591.1 | - | 1415 | |||
| AK052040.1 | - | 2, 3 | 342 | |||
| AY351699.1 | - | 2, 3 | 317 | |||
| BC038036.1 | - | 2, 3 | 595 | |||

Sdk2
Genomic Map