siRNA Details
| Gene Symbol: Uba6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172712.2 | NP_766300.1 | 4, 5 | 271 | ||
| GenBank mRNA | AK029769.1 | - | 4, 5 | 301 | ||
| AK049603.1 | - | 4, 5 | 309 | |||
| AK053583.1 | - | 4, 5 | 268 | |||
| AK086539.1 | - | 4, 5 | 289 | |||
| BC063048.1 | - | 4, 5 | 271 | 6825308 | ||

Uba6
Genomic Map