siRNA Details
| Gene Symbol: Fbxw8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172721.2 | NP_766309.2 | 3 | 526 | ||
| GenBank mRNA | AK031815.1 | - | 3 | 520 | ||
| AK034447.1 | - | 3 | 454 | |||
| AK047695.1 | - | 3 | 494 | |||
| AK047756.1 | - | 3 | 468 | |||
| AK053794.1 | - | 3 | 527 | |||
| AK134291.1 | - | 3 | 533 | |||
| AK194533.1 | - | 2 | 118 | |||
| BC009095.1 | - | 3 | 479 | 3158218 | ||

Fbxw8
Genomic Map