siRNA Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172723.4 | NP_766311.2 | 4, 5 | 482 | ||
| GenBank mRNA | AK076754.1 | - | 3, 4 | 468 | ||
| AK076768.1 | - | 4, 5 | 239 | |||
| BC056934.1 | - | 3, 4 | 208 | |||
| BC079634.1 | - | 3, 4 | 276 | |||
| BC144874.1 | - | 3, 4 | 227 | |||

Adap1
Genomic Map