siRNA Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172772.2 | NP_766360.2 | 3 | 985 | ||
| GenBank mRNA | AK035930.1 | - | 3 | 986 | ||
| AK046403.1 | - | 3 | 972 | |||
| AK046623.1 | - | 3 | 959 | |||
| AK220158.1 | - | 3 | 1014 | |||
| BC049357.1 | - | 3 | 962 | |||
| BC055816.1 | - | 3 | 713 | |||
| BC058683.1 | - | 3 | 943 | 6834871 | ||
| BC069845.1 | - | 3 | 698 | |||

Fam63b
Genomic Map