siRNA Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161430.2 | NP_001154902.1 | 5 | 592 | ||
| NM_001290532.1 | NP_001277461.1 | 5 | 677 | |||
| NM_001290533.1 | NP_001277462.1 | 4 | 683 | |||
| NM_172782.4 | NP_766370.2 | 4 | 629 | |||
| GenBank mRNA | AK032102.1 | - | 4 | 600 | ||
| AK144349.1 | - | 4 | 613 | |||
| BC064727.1 | - | 4 | 604 | |||
| BC068166.1 | - | 4 | 582 | 30361776 | ||
| BC094570.1 | - | 4 | 546 | 30361291 | ||
| BC118503.1 | - | 4 | 557 | 40106658 | ||
| CA482101.1 | - | 5 | 628 | |||

Nxt2
Genomic Map