siRNA Details
| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 5 | 645 | ||
| GenBank mRNA | AK030591.1 | - | 1743 | |||
| AK052040.1 | - | 5 | 670 | |||
| AK129379.1 | - | 1 | 26 | |||
| AY351699.1 | - | 5 | 645 | |||
| BC038036.1 | - | 5 | 923 | |||

Sdk2
Genomic Map