siRNA Details
| Gene Symbol: Syt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172804.2 | NP_766392.2 | 5 | 950 | ||
| GenBank mRNA | AB102950.1 | - | 4 | 1282 | ||
| AK032606.1 | - | 6 | 1729 | |||
| AK137129.1 | - | 5 | 958 | |||
| BC056932.1 | - | 4 | 1060 | 5708126 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008764729, XM_234300, XM_008764730.

Syt16
Genomic Map