siRNA Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172711.2 | NP_766299.1 | 6, 7 | 724 | ||
| GenBank mRNA | AK032597.1 | - | 5, 6 | 1732 | ||
| AK047411.1 | - | 6, 7 | 593 | |||
| AK053420.1 | - | 6, 7 | 698 | |||
| AK084627.1 | - | 6, 7 | 705 | |||
| AK140534.1 | - | 6, 7 | 683 | |||
| AK160956.1 | - | 6, 7 | 743 | |||
| AK165012.1 | - | 6, 7 | 713 | |||
| BC057962.1 | - | 6, 7 | 678 | 5353891 | ||

Guf1
Genomic Map