siRNA Details
| Gene Symbol: SIGLEC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011527533.2 | XP_011525835.1 | 2 | 554 | ||
| XM_011527534.2 | XP_011525836.1 | 2 | 557 | |||
| XM_011527535.2 | XP_011525837.1 | 2 | 501 | |||
| XM_011527536.2 | XP_011525838.1 | 2 | 393 | |||
| XM_011527537.2 | XP_011525839.1 | 2 | 393 | |||
| XM_011527538.2 | XP_011525840.1 | 2 | 556 | |||
| XM_011527539.2 | XP_011525841.1 | 2 | 555 | |||
| XM_017027511.1 | XP_016883000.1 | 2 | 557 | |||
| GenBank mRNA | AK300170.1 | - | 2 | 394 | ||
| AK300182.1 | - | 2 | 492 | |||
| AK308174.1 | - | 2 | 501 | |||
| AK309903.1 | - | 2 | 393 | |||
| AU138048.1 | - | 2 | 501 | |||
| BC035359.1 | - | 2 | 415 | 5176194 | ||
| BC140798.1 | - | 2 | 365 | 9021666 | ||
| CA495365.1 | - | 1 | 100 | |||
| D86358.1 | - | 2 | 549 | |||
| D86359.1 | - | 2 | 549 | |||
| U71382.1 | - | 2 | 349 | |||

SIGLEC6
Genomic Map