siRNA Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172723.4 | NP_766311.2 | 9 | 952 | ||
| GenBank mRNA | AK076754.1 | - | 8 | 938 | ||
| AK076768.1 | - | 9 | 709 | |||
| BC056934.1 | - | 8 | 678 | |||
| BC079634.1 | - | 8 | 746 | |||
| BC144874.1 | - | 8 | 694 | |||

Adap1
Genomic Map