siRNA Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172727.3 | NP_766315.1 | 6 | 976 | ||
| GenBank mRNA | AK052323.1 | - | 6 | 890 | ||
| AK140239.1 | - | 6 | 882 | |||
| BC039996.1 | - | 3 | 303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam221a| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172727.3 | NP_766315.1 | 6 | 976 | ||
| GenBank mRNA | AK052323.1 | - | 6 | 890 | ||
| AK140239.1 | - | 6 | 882 | |||
| BC039996.1 | - | 3 | 303 | |||