siRNA Details
| Gene Symbol: Zfp954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172738.3 | NP_766326.2 | 5 | 1078 | ||
| GenBank mRNA | AK033116.1 | - | 5 | 1054 | ||
| BC055688.1 | - | 5 | 1047 | 6402770 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp954| Gene Symbol: Zfp954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172738.3 | NP_766326.2 | 5 | 1078 | ||
| GenBank mRNA | AK033116.1 | - | 5 | 1054 | ||
| BC055688.1 | - | 5 | 1047 | 6402770 | ||