siRNA Details
| Gene Symbol: Plekha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040271.1 | - | 13, 14 | 1979 | ||
| AK146866.1 | - | 2170 | ||||
| AK160810.1 | - | 13, 14 | 2059 | |||
| BC060678.1 | - | 12, 13 | 1900 | |||
| BC084587.1 | - | 13, 14 | 2074 | 3602481 | ||
| EU380771.1 | - | 2196 | ||||
| EU380772.1 | - | 2254 | ||||

Plekha7
Genomic Map