siRNA Details
| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172748.2 | NP_766336.2 | 11 | 2514 | ||
| GenBank mRNA | AK028405.1 | - | 11 | 2281 | ||
| AK031431.1 | - | 9 | 1907 | |||
| AK161937.1 | - | 2 | 485 | |||
| AK162161.1 | - | 5 | 893 | |||
| BC053109.1 | - | 7 | 1507 | |||
| BC059812.1 | - | 11 | 2514 | 6401846 | ||

Fbxl19
Genomic Map