siRNA Details
| Gene Symbol: Adprhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172750.3 | NP_766338.1 | 5 | 838 | ||
| GenBank mRNA | AJ427360.1 | - | 5 | 815 | ||
| AK081786.1 | - | 5 | 672 | |||
| AK084501.1 | - | 5 | 839 | |||
| BC063759.1 | - | 5 | 828 | 30134000 | ||

Adprhl1
Genomic Map