siRNA Details
| Gene Symbol: Heatr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172757.3 | NP_766345.3 | 7 | 923 | ||
| GenBank mRNA | AK047782.1 | - | 7 | 954 | ||
| AK052635.1 | - | 7 | 950 | |||
| AK134510.1 | - | 7 | 928 | |||
| AK136424.1 | - | 7 | 933 | |||
| AK140522.1 | - | 7 | 941 | |||
| AK163715.1 | - | 7 | 910 | |||
| AK165541.1 | - | 7 | 817 | |||
| BC030373.1 | - | 3 | 268 | |||
| BC065060.1 | - | 7 | 923 | 6402217 | ||

Heatr3
Genomic Map