siRNA Details
| Gene Symbol: Nfrkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172766.3 | NP_766354.2 | 19, 20 | 2177 | ||
| GenBank mRNA | AK041206.1 | - | 19, 20 | 2415 | ||
| AK049804.1 | - | 5, 6 | 552 | |||
| BC033595.1 | - | 19, 20 | 2167 | 4947357 | ||
| BC059861.1 | - | 19, 20 | 2224 | |||
| BC059900.1 | - | 10, 11 | 1824 | |||
| BC060076.1 | - | 19, 20 | 2406 | |||

Nfrkb
Genomic Map