siRNA Details
| Gene Symbol: Gramd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172768.1 | NP_766356.1 | 20 | 2655 | ||
| GenBank mRNA | AK028277.1 | - | 21 | 2550 | ||
| AK033156.1 | - | 20 | 2655 | |||
| AK122465.1 | - | 20 | 2627 | |||
| BC030911.1 | - | 11 | 1399 | |||
| BC070451.1 | - | 20 | 2501 | 30616509 | ||

Gramd1b
Genomic Map