siRNA Details
| Gene Symbol: Slc22a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172776.2 | NP_766364.1 | 10 | 1823 | ||
| GenBank mRNA | AF536195.1 | - | 10 | 1821 | ||
| AF536196.1 | - | 11 | 1631 | |||
| AK052597.1 | - | 10 | 1824 | |||
| BC126952.1 | - | 11 | 1562 | 40090863 | ||
| Gene Symbol: Slc22a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF408394.1 | - | 1579 | |||

Slc22a29
Genomic Map