siRNA Details
| Gene Symbol: CDC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253.3 | NP_001244.1 | 3 | 522 | ||
| GenBank mRNA | AB007892.2 | - | 3 | 294 | ||
| AK128737.1 | - | 3 | 330 | |||
| AK299738.1 | - | 3 | 330 | |||
| BC001568.1 | - | 3 | 311 | 3452352 | ||
| BM829346.1 | - | 3 | 522 | |||
| D85423.1 | - | 2 | 191 | |||
| DC422730.1 | - | 3 | 521 | |||
| U86753.1 | - | 3 | 212 | |||

CDC5L
Genomic Map