siRNA Details
| Gene Symbol: Syt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172804.2 | NP_766392.2 | 7 | 2482 | ||
| GenBank mRNA | AB102950.1 | - | 6 | 2814 | ||
| AK032606.1 | - | 8 | 3261 | |||
| AK138302.1 | - | 676 | ||||
| AV348245.2 | - | 1 | 452 | |||
| BC056932.1 | - | 6 | 2592 | 5708126 | ||

Syt16
Genomic Map