siRNA Details
| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172806.2 | NP_766394.2 | 4 | 1088 | ||
| GenBank mRNA | AK033160.1 | - | 3 | 1001 | ||
| AK122522.1 | - | 2 | 738 | |||
| AK134271.1 | - | 3 | 939 | |||
| BC041669.1 | - | 4 | 1088 | 3482657 | ||
| BC057303.1 | - | 3 | 807 | 5702276 | ||

Btbd7
Genomic Map