siRNA Details
Gene Symbol: Lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172814.3 | NP_766402.1 | 4 | 534 | ||
GenBank mRNA | AK050512.1 | - | 4 | 535 | ||
BC058345.1 | - | 3 | 714 | 6822657 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172814.3 | NP_766402.1 | 4 | 534 | ||
GenBank mRNA | AK050512.1 | - | 4 | 535 | ||
BC058345.1 | - | 3 | 714 | 6822657 |