siRNA Details
| Gene Symbol: Slc30a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172816.3 | NP_766404.1 | 2 | 483 | ||
| GenBank mRNA | AK050482.1 | - | 2 | 384 | ||
| AK050494.1 | - | 2 | 483 | |||
| BC117544.1 | - | 2 | 315 | 40105617 | ||
| BC118521.1 | - | 2 | 312 | 40105622 | ||

Slc30a8
Genomic Map