siRNA Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172852.3 | NP_766440.2 | 2, 3 | 236 | ||
| GenBank mRNA | AK030670.1 | - | 2, 3 | 238 | ||
| BC117990.1 | - | 2, 3 | 226 | 40104505 | ||
| BC117991.1 | - | 2, 3 | 226 | |||

Serpinb13
Genomic Map