siRNA Details
| Gene Symbol: Zfp462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172867.3 | NP_766455.2 | 8 | 6851 | ||
| GenBank mRNA | AK031378.1 | - | 3 | 362 | ||
| DQ355518.1 | - | 7 | 6548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp462| Gene Symbol: Zfp462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172867.3 | NP_766455.2 | 8 | 6851 | ||
| GenBank mRNA | AK031378.1 | - | 3 | 362 | ||
| DQ355518.1 | - | 7 | 6548 | |||