siRNA Details
Gene Symbol: Ttc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172878.3 | NP_766466.3 | 4 | 784 | ||
GenBank mRNA | AK030055.1 | - | 4 | 758 | ||
BC068299.1 | - | 636 | 30094917 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172878.3 | NP_766466.3 | 4 | 784 | ||
GenBank mRNA | AK030055.1 | - | 4 | 758 | ||
BC068299.1 | - | 636 | 30094917 |