siRNA Details
| Gene Symbol: Tmem132d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172885.2 | NP_766473.1 | 2 | 1045 | ||
| GenBank mRNA | AB100355.1 | - | 2 | 499 | ||
| AK083210.1 | - | 2 | 1046 | |||
| BC140953.1 | - | 2 | 585 | 9055565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem132d| Gene Symbol: Tmem132d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172885.2 | NP_766473.1 | 2 | 1045 | ||
| GenBank mRNA | AB100355.1 | - | 2 | 499 | ||
| AK083210.1 | - | 2 | 1046 | |||
| BC140953.1 | - | 2 | 585 | 9055565 | ||