siRNA Details
| Gene Symbol: Svs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172888.3 | NP_766476.2 | 1 | 465 | ||
| GenBank mRNA | AK035222.1 | - | 1 | 466 | ||
| AY115568.1 | - | 1 | 422 | |||
| AY283179.1 | - | 1 | 422 | |||
| BC125635.1 | - | 1 | 336 | 40130150 | ||

Svs1
Genomic Map