siRNA Details
| Gene Symbol: Nlrp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172896.2 | NP_766484.1 | 3 | 393 | ||
| GenBank mRNA | AK054378.1 | - | 3 | 283 | ||
| AK087843.1 | - | 3 | 394 | |||
| AK136205.1 | - | 4 | 369 | |||
| AK166640.1 | - | 4 | 369 | |||
| AY355341.1 | - | 3 | 394 | |||
| AY596200.1 | - | 1 | 90 | |||
| BC141342.1 | - | 3 | 293 | 9055954 | ||
| BC141343.1 | - | 3 | 293 | 9055955 | ||

Nlrp4a
Genomic Map