siRNA Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172815.3 | NP_766403.1 | 3 | 1083 | ||
| GenBank mRNA | AK049891.1 | - | 3 | 834 | ||
| AK052457.1 | - | 2 | 1207 | |||
| AK087485.1 | - | 3 | 1084 | |||
| BC052844.1 | - | 3 | 434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rspo2| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172815.3 | NP_766403.1 | 3 | 1083 | ||
| GenBank mRNA | AK049891.1 | - | 3 | 834 | ||
| AK052457.1 | - | 2 | 1207 | |||
| AK087485.1 | - | 3 | 1084 | |||
| BC052844.1 | - | 3 | 434 | |||