siRNA Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172857.2 | NP_766445.1 | 6, 7 | 1163 | ||
| GenBank mRNA | AK030132.1 | - | 6, 7 | 1165 | ||
| BC092241.1 | - | 4, 5 | 417 | 30670567 | ||
| BC144750.1 | - | 5, 6 | 546 | 9053298 | ||
| BC150662.1 | - | 6, 7 | 640 | 9087573 | ||

Exd1
Genomic Map