siRNA Details
| Gene Symbol: Mfsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172883.2 | NP_766471.1 | 5, 6 | 759 | ||
| GenBank mRNA | AK029021.1 | - | 5, 6 | 759 | ||
| AK087226.1 | - | 5, 6 | 969 | |||
| AK171069.1 | - | 4, 5 | 3391 | |||
| BC094512.1 | - | 3, 4 | 371 | 2616131 | ||

Mfsd7a
Genomic Map