siRNA Details
| Gene Symbol: 2900026A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172884.3 | NP_766472.1 | 3 | 600 | ||
| GenBank mRNA | AK041243.1 | - | 3 | 579 | ||
| AK154484.1 | - | 3 | 639 | |||
| AK171617.1 | - | 3 | 600 | |||
| AK220411.1 | - | 1 | 46 | |||
| BC147338.1 | - | 3 | 479 | 9056390 | ||
| BC147339.1 | - | 3 | 479 | 9056416 | ||

2900026A02Rik
Genomic Map