siRNA Details
| Gene Symbol: Slc30a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172816.3 | NP_766404.1 | 8 | 1828 | ||
| GenBank mRNA | AK050482.1 | - | 8 | 1729 | ||
| AK050494.1 | - | 8 | 1828 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a8| Gene Symbol: Slc30a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172816.3 | NP_766404.1 | 8 | 1828 | ||
| GenBank mRNA | AK050482.1 | - | 8 | 1729 | ||
| AK050494.1 | - | 8 | 1828 | |||