siRNA Details
| Gene Symbol: ACTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104.3 | NP_001095.2 | 3 | 394 | ||
| NM_001258371.2 | NP_001245300.2 | 3 | 542 | |||
| NR_047663.2 | 3 | 394 | ||||
| GenBank mRNA | AK125851.1 | - | 3 | 928 | ||
| AK303044.1 | - | 3 | 542 | |||
| AK313556.1 | - | 3 | 332 | |||
| BC099647.3 | - | 3 | 315 | 40008220 | ||
| BC099649.3 | - | 3 | 315 | 40008223 | ||
| DA915904.1 | - | 3 | 396 | |||
| M86407.1 | - | 3 | 315 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013456, AF093775, BC111890, BY132390.

ACTN3
Genomic Map