siRNA Details
| Gene Symbol: ACYP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107.4 | NP_001098.1 | 3 | 286 | ||
| NM_001302616.1 | NP_001289545.1 | 3 | 268 | |||
| NM_001302617.1 | NP_001289546.1 | 3 | 464 | |||
| NR_126393.1 | 4 | 365 | ||||
| NR_126394.1 | 2 | 372 | ||||
| NR_126395.1 | 2 | 194 | ||||
| GenBank mRNA | AK312101.1 | - | 3 | 286 | ||
| BC035568.1 | - | 3 | 271 | 5392248 | ||
| BG614847.1 | - | 4 | 332 | |||
| BP316920.1 | - | 3 | 249 | |||
| BX104912.1 | - | 2 | 139 | |||
| CD675334.1 | - | 3 | 473 | |||
| DB071115.1 | - | 3 | 286 | |||
| HY012032.1 | - | 2 | 334 | |||
| X84194.1 | - | 2 | 250 | |||

ACYP1
Genomic Map